Skip to Content
Merck
All Photos(1)

Documents

05-23-2401

Sigma-Aldrich

Calcitonin, Salmon

Calcitonin, Salmon, CAS 47931-85-1, is a 32 amino acid synthetic calcitonin that is shown to stimulate bone formation and inhibit bone resorption. Has ability to cross mucous membranes.

Synonym(s):

Calcitonin, Salmon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C145H240N44O48S2
CAS Number:
Molecular Weight:
3431.85
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

sterility

non-sterile

Assay

≥98% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

5% acetic acid: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C145H240N44O48S2/c1-65(2)45-86(175-139(232)110(70(11)12)183-136(229)99-63-239-238-62-79(148)117(210)178-96(59-191)134(227)174-92(52-104(151)202)131(224)172-90(49-69(9)10)129(222)180-98(61-193)135(228)187-114(74(16)197)142(235)181-99)118(211)158-55-106(204)162-80(25-18-20-40-146)120(213)169-89(48-68(7)8)128(221)179-97(60-192)133(226)167-83(34-37-102(149)200)122(215)165-85(36-39-109(207)208)123(216)171-88(47-67(5)6)127(220)173-91(51-77-54-156-64-161-77)130(223)164-81(26-19-21-41-147)121(214)170-87(46-66(3)4)126(219)166-84(35-38-103(150)201)125(218)186-113(73(15)196)141(234)177-94(50-76-30-32-78(199)33-31-76)143(236)189-44-24-29-101(189)137(230)168-82(27-22-42-157-145(154)155)124(217)185-112(72(14)195)140(233)176-93(53-105(152)203)132(225)184-111(71(13)194)138(231)160-56-107(205)163-95(58-190)119(212)159-57-108(206)182-115(75(17)198)144(237)188-43-23-28-100(188)116(153)209/h30-33,54,64-75,79-101,110-115,190-199H,18-29,34-53,55-63,146-148H2,1-17H3,(H2,149,200)(H2,150,201)(H2,151,202)(H2,152,203)(H2,153,209)(H,156,161)(H,158,211)(H,159,212)(H,160,231)(H,162,204)(H,163,205)(H,164,223)(H,165,215)(H,166,219)(H,167,226)(H,168,230)(H,169,213)(H,170,214)(H,171,216)(H,172,224)(H,173,220)(H,174,227)(H,175,232)(H,176,233)(H,177,234)(H,178,210)(H,179,221)(H,180,222)(H,181,235)(H,182,206)(H,183,229)(H,184,225)(H,185,217)(H,186,218)(H,187,228)(H,207,208)(H4,154,155,157)

InChI key

BBBFJLBPOGFECG-UHFFFAOYSA-N

Related Categories

General description

Synthetic, salmon calcitonin. A 32 amino acid residue peptide that is shown to stimulate bone formation and inhibit bone resorption. Has ability to cross mucous membranes. Has no significant effect on inositol phosphate accumulation.
Synthetic, salmon calcitonin. Calcitonin is a single-chain peptide hormone of 32 amino acid residues with a disulfide bridge between cysteines 1 and 7. It is produced in the parafollicular C cells of the thyroid gland and acts to stimulate bone formation by osteoblasts and inhibits bone resorption. Part of the effect of calcitonin appears to be mediated through increasing cAMP concentrations in osteoclasts. Has ability to cross mucus membranes. Has no significant effect on inositol phosphate accumulation.

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Cys¹-Ser-Asn-Leu-Ser-Thr-Cys⁷-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH₂(disulfide bond: 1 → 7)

Physical form

Sold on the basis of peptide content.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Gennari, C. and Agnusdei, D. 1994. Br. J. Clin. Pract.48, 196.
Reginster, J.Y. 1993. Osteoporos. Int. 3 (Suppl. 2), S3.
MacIntyre, I. et al. 1988. Clin. Orthop.217, 45.
Chambers, T.J., et al. 1985. J. Pathol.145, 297.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service