Skip to Content
Merck
All Photos(4)

Documents

05-1951

Sigma-Aldrich

Anti-trimethyl Histone H3 (Lys27) Antibody, clone 18E9.1

clone 18E9.1, from mouse

Synonym(s):

H3K27me3, Histone H3 (tri methyl K27), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody product type

primary antibodies

clone

18E9.1, monoclonal

species reactivity

human, mouse

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys27)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

This antibody recognizes Histone H3 trimethylated at Lys27.

Immunogen

Epitope: Trimethylated Lys27
Linear peptide corresponding to human Histone H3 trimethylated at Lys27.

Application

Anti-trimethyl Histone H3 (Lys27) Antibody, clone 18E9.1 is a mouse monoclonal antibody for detection of trimethyl Histone H3 (Lys27) also known as H3K27me3, Histone H3 (tri methyl K27) & has been validated in WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blot Analysis: A 1:25,000 dilution from a representative lot detected Histone H3 in 10 µg of modified and nonmodified peptides.

Western Blot (Snap i.d.) Analysis: A 1:25,000 dilution from a representative lot detected Histone H3 in 10 µg of HeLa acid extract.

Immunocytochemistry Analysis: A 1:125 dilution from a representative lot detected Histone H3 in A431 and NIH/3T3 cells.

Dot Blot Analysis: Histone peptides with various modifications (see table) were transferred to
PVDF membrane and probed with Anti-Trimethyl Histone H3 (Lys4) (1:100 dilution). Proteins were visualized using a Donkey Anti-rabbit IgG conjugated to HRP and a chemiluminescence detection systems.

Quality

Evaluated by Western Blot in HeLa acid extract.

Western Blot Analysis: A 1:50,000 dilution of this antibody detected Histone H3 in 10 µg of HeLa acid extract.

Target description

~17 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service