Skip to Content
Merck
All Photos(1)

Key Documents

05-180-I

Sigma-Aldrich

Anti-Cortactin (p80/85) Antibody, clone 4F11

clone 4F11

Synonym(s):

Anti-Cortactin antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CHF 160.00
100 μL
CHF 408.00

CHF 160.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
CHF 160.00
100 μL
CHF 408.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 160.00


Estimated to ship on25 May 2025


Request a Bulk Order

clone

4F11, monoclonal

Quality Level

form

liquid

isotype

IgG1κ

target post-translational modification

unmodified

Specificity

Clone 4F11 is a mouse monoclonal antibody that detects Cortactin.

Immunogen

Mixture of affinity-purified tyrosine phospho-proteins from chick embryo fibroblasts expressing activated pp60src

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service