Skip to Content
Merck
All Photos(1)

Documents

05-1533

Sigma-Aldrich

Anti-Gemin3 Antibody, clone 12H12

clone 12H12, from mouse

Synonym(s):

Component of gems 3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 20, DEAD box protein 20, DEAD box protein DP 103, DEAD-box protein DP103, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD, SMN-interacting protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12H12, monoclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DDX20(11218)

General description

Gemin3 is a member of the DEAD box protein family. The members of this family are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD). Some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Gemin 3 is a component of the survival of motor neurons (SMN) complex and codes for a DEAD box protein with an ATPase activity The SMN complex is essential for the biogenesis of spliceosomal small nuclear ribonucleoproteins and likely functions in the assembly, metabolism, and transport of a diverse number of other ribonucleoproteins. Specifically, the SMN complex assembles 7 Sm proteins (Gemin2-8) into a core structure around a highly conserved sequence of ribonucleic acid (RNA) found in small nuclear RNAs. The complex recognizes specific sequences and structural features of small nuclear RNAs and Sm proteins and assembles small nuclear ribonucleoproteins in a stepwise fashion.

Specificity

This antibody recognizes Gemin-3.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human Gemin-3.

Application

Anti-Gemin3 Antibody, clone 12H12 is a Mouse Monoclonal Antibody for detection of Gemin3 also known as Component of gems 3, DEAD-box protein DP103 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected Gemin-3 on 10 µg of HeLa cell lysate.

Target description

Detects a band of approximately 105 kDa (Calculated molecular weight: 92 kDa)

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgGκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sukhleen Kour et al.
Nature communications, 12(1), 2558-2558 (2021-05-09)
GEMIN5, an RNA-binding protein is essential for assembly of the survival motor neuron (SMN) protein complex and facilitates the formation of small nuclear ribonucleoproteins (snRNPs), the building blocks of spliceosomes. Here, we have identified 30 affected individuals from 22 unrelated
Anne-Sophie Gribling-Burrer et al.
Nucleic acids research, 45(9), 5399-5413 (2017-01-25)
Selenoprotein synthesis requires the co-translational recoding of a UGASec codon. This process involves an RNA structural element, called Selenocysteine Insertion Sequence (SECIS) and the SECIS binding protein 2 (SBP2). Several selenoprotein mRNAs undergo unusual cap hypermethylation by the trimethylguanosine synthase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service