Skip to Content
Merck
All Photos(1)

Key Documents

04-795

Sigma-Aldrich

Anti-phospho-Ribosomal Protein S6 (Ser235) Antibody, clone MC27, rabbit monoclonal

culture supernatant, clone MC27, Upstate®

Synonym(s):

40S ribosomal protein S6, Phosphoprotein NP33, ribosomal protein S6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 282.00

CHF 282.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 282.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 282.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MC27, monoclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer235)

Gene Information

human ... RPS6(6194)

General description

S6 (Ribosomal protein S6, RpS6, 40S ribosomal protein S6) is known to play a role in cell growth and proliferation. It lays downstream of both the PI3K/Akt/mTOR pathway as a substrate of p70 S6 kinase and the Ras/Erk pathway as a substrate of RSK. S6 is phosphorylated in response to stimulation by growth factors, tumor promoting agents, and mitogens. It is dephosphorylated at growth arrest. There are two major areas of S6 phosphorylation; Ser235/Ser236 that is known to be phosphorylated by both p70S6 Kinase as well as RSK as well as Ser240/244 which is thought to only be phosphorylated by p70S6K.

Specificity

Broad species cross-reactivity predicted, due to sequence homology.
Recognizes ribosomal protein S6 only when phosphorylated on Ser235.

Immunogen

Epitope: Phosphorylated Ser235
Synthetic peptide containing the sequence …RRL[pS]SL… in which pS corresponds to phospho-serine at residue 235 of human ribosomal protein S6. The immunizing sequence is identical in mouse, rat, chicken and Xenopus.

Application

Anti-phospho-Ribosomal Protein S6 (Ser235) Antibody, clone MC27 is an antibody against phospho-Ribosomal Protein S6 (Ser235) for use in WB.
Research Category
Signaling

Apoptosis & Cancer
Research Sub Category
PI3K, Akt, & mTOR Signaling
Western Blot Analysis:
A 1:500-1:2,000 dilution of this lot detected phosphorylated ribosomal protein S6 in lysates from serum stimulated 3T3 cells.

Quality

Routinely evaluated by western blot on serum stimulated 3T3 cell lysate.

Target description

~30 kDa

Physical form

Cultured supernatant with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Serum stimulated 3T3 cell lysate.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service