Skip to Content
Merck
All Photos(1)

Documents

03-308

Sigma-Aldrich

Magna ChIRP® NEAT1 lncRNA Probe Set

The Magna ChIRP NEAT1 lncRNA Probe Set contains 33 predesigned 20-mer DNA oligonucleotides tiled along and complementary to the sequence of human lncRNA NEAT1.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.52

Quality Level

manufacturer/tradename

Magna ChIRP®

technique(s)

DNA sequencing: suitable
RNA extraction: suitable (nuclear function kit)
RNA purification: suitable (with magnetic beads)
activity assay: suitable (protein interaction)
nucleic acid detection: suitable (RNA)
protein purification: suitable (protein determination)

shipped in

dry ice

Related Categories

General description

ChIRP (Chromatin Isolation by RNA Purification), allows recovery of specific segments of DNA and proteins using chromatin associated RNA as a target for biotinylated antisense probes. The Magna ChIRP NEAT1 lncRNA Probe Set contains 33 predesigned 20-mer DNA oligonucleotides tiled along and complementary to the sequence of human lncRNA NEAT1. All probes are biotin-TEG-labeled at the 3-prime end. The oligonucleotides are split to two pools (even and odd) and are supplied ready for use in the ChIRP method. This probe set is fully compatible with the Maga ChIRP kits (cat. # 17-10494 and 17-10495).

Application

Research Category
Epigenetics & Nuclear Function
The Magna ChIRP NEAT1 lncRNA Probe Set contains 33 predesigned 20-mer DNA oligonucleotides tiled along and complementary to the sequence of human lncRNA NEAT1.

Physical form

50 µM biotin-TEG-labeled 20-mer DNA oligonucleotides in containing 10 mM Tris-HCl, 1mM EDTA, pH 8.0

Storage and Stability

Upon receipt store at -20°C. Avoid multiple freeze thaws. With proper storage and handling this product is stable for 6 months from the date of receipt.

Legal Information

MAGNA CHIRP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service