Skip to Content
Merck
All Photos(1)

Key Documents

03-0205-00

Millipore

SMC® Human TDP-43 Immunoassay Kit

kit sufficient for 96 wells

Synonym(s):

TDP-43 Single Molecule Counting kit, TDP-43 immunoassay kit, TDP-43 ultra-sensitive immunoassay, Transactive Response DNA-binding Protein 43 immunoassay kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

usage

 kit sufficient for 96 wells

species reactivity

human

packaging

pkg of 1 ea

manufacturer/tradename

SMC®

parameter

100 μL sample volume (CSF)
12.5 μL sample volume (serum and plasma)

assay range

intra-assay cv: ≤7%
(n=5)

linearity: 80-120%
recovery range: 80-120%
sensitivity: 5.21 pg/mL
standard curve range: 2.6-6,000 pg/mL
inter-assay cv: ≤11%
(n=3)

technique(s)

single molecule counting: suitable

input

cerebrospinal fluid (CSF)
plasma
serum

shipped in

wet ice

storage temp.

2-8°C

General description

The human Transactive Response DNA-binding Protein 43 (TDP-43) is a transcriptional repressor which can bind to both DNA and RNA, and regulate functions such as mRNA splicing, microRNA biogenesis, and transcription. While this 414 amino acid long protein can be found in a variety of tissue types, it is most frequently studied in the context of neuroscience research, as misfolding, phosphorylation, and aggregation of TDP-43 in neurons can has been observed in a variety of neurodegenerative disorders, including frontotemporal dementia (FTL), amyotrophic lateral sclerosis (ALS), and certain subsets of Alzheimer’s disease patients. To empower research to evaluate TDP-43 as a cognitive biomarker in both healthy and disease states such as ALS and FTL, the new SMC® Human TDP-43 High Sensitivity Immunoassay Kit scientists can measure this analyte in CSF, serum, and plasma samples even at low abundance.

Research Category:
Neuroscience
neurological

Research Sub-Category:
Alzheimer′s Disease
Aging
Nervous System
Neurodegenerative Diseases
Neurofilament & Neuron Metabolism
Neuronal & Glial Markers
Neuroregenerative Medicine
Neuroscience

Technique(s)
SMC® : High sensitivity protein biomarker detection immunoassay

Specificity

This assay was tested for cross-reactivity with phospho-α-synuclein (S129) and phospho-Tau (T181). No cross-reactivity was detected with these analytes.

Application

used for ultrasensitive quantification of Human TDP-43 in human serum, plasma, and CSF.

Features and Benefits

This SMC® kit features:
  • Accurate and reproducible performance on the state-of-the-art SMCxPRO® ultrasensitive immunoassay system
  • Compatibility with automated liquid handling

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Discover how to expand neuroscience biomarker research with high sensitivity immunoassays, such as Single Molecule Counting (SMC®) assays. These assays give researchers the tools they need to detect low-abundant blood-based biomarkers and expand biomarker options in neuroscience research.

Explore high-sensitivity SMC® neurology immunoassays that precisely measure low levels of neurology protein analytes, such as NF-L or Tau proteins.

Explore ultrasensitive immunoassay technology with SMC® technology, including its biomarker detection platform, advantages of high sensitivity assays, and more.

Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service