Skip to Content
Merck
All Photos(1)

Key Documents

ENAH97BE73A3

Enamine

3-aminobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
121.14
MDL number:

Iupac Name

3-aminobenzaldehyde

Inchi Code

1S/C7H7NO/c8-7-3-1-2-6(4-7)5-9/h1-5H,8H2

InChI key

SIXYIEWSUKAOEN-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-263597

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library