ENAH38A2B967
Enamine
4-cyclopropyl-2-fluorobenzene-1-sulfonyl fluoride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-cyclopropyl-2-fluorobenzenesulfonyl fluoride
Inchi Code
1S/C9H8F2O2S/c10-8-5-7(6-1-2-6)3-4-9(8)14(11,12)13/h3-6H,1-2H2
InChI key
TWVXXBXJMRPTAY-UHFFFAOYSA-N
storage temp.
RT
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-6485964
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://www.enaminestore.com/catalog/EN300-6485964
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.