Iupac Name
4-(trichloromethyl)tetrahydro-2H-pyran-4-ol
Inchi Code
1S/C6H9Cl3O2/c7-6(8,9)5(10)1-3-11-4-2-5/h10H,1-4H2
InChI key
HWJVEYYSLHKMCN-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-6496241
MSDS
https://www.enaminestore.com/catalog/EN300-6496241
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.