ENA371464594
Enamine
1-[4-(pyridin-1-ium-1-yl)butyl]pyridin-1-ium dibromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-[4-(1-pyridiniumyl)butyl]pyridinium dibromide
Inchi Code
1S/C14H18N2.2BrH/c1-3-9-15(10-4-1)13-7-8-14-16-11-5-2-6-12-16;;/h1-6,9-12H,7-8,13-14H2;2*1H/q+2;;/p-2
InChI key
OZXBTPBFHBTLJM-UHFFFAOYSA-L
Salt Data
[Br-]
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-111176
MSDS
https://www.enaminestore.com/catalog/EN300-111176
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.