COMH97F0E44E
Combi-Blocks, Inc.
[1-(3-Bromobenzyl)-1h-imidazol-5-yl]methanamine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(1-(3-bromobenzyl)-1H-imidazol-5-yl)methanamine hydrochloride
Inchi Code
1S/C11H12BrN3.ClH/c12-10-3-1-2-9(4-10)7-15-8-14-6-11(15)5-13;/h1-4,6,8H,5,7,13H2;1H
InChI key
GIRHPKIZJSIHNU-UHFFFAOYSA-N
Purity
99%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?JN-6971
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?JN-6971
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.