COMH4211F488
Combi-Blocks, Inc.
N-ethyl-2-(4-nitrophenoxy)acetamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N-ethyl-2-(4-nitrophenoxy)acetamide
Inchi Code
1S/C10H12N2O4/c1-2-11-10(13)7-16-9-5-3-8(4-6-9)12(14)15/h3-6H,2,7H2,1H3,(H,11,13)
InChI key
RDXDZQDKGIZBCH-UHFFFAOYSA-N
Purity
97%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?QL-4850
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QL-4850
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.