COMH04237D80
Combi-Blocks, Inc.
Sodium 4-methylpiperazine-1-carbodithioate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
sodium 4-methyl-1-piperazinecarbodithioate
Inchi Code
1S/C6H12N2S2.Na/c1-7-2-4-8(5-3-7)6(9)10;/h2-5H2,1H3,(H,9,10);/q;+1/p-1
InChI key
JPKVEDOCKBMPHK-UHFFFAOYSA-M
Purity
95%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3309
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3309
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.