C12H316CF307
Chemcia Scientific, LLC
4-Bromo-3H-isobenzofuran-1-ylideneamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-bromoisobenzofuran-1(3H)-imine
Inchi Code
1S/C8H6BrNO/c9-7-3-1-2-5-6(7)4-11-8(5)10/h1-3,10H,4H2
InChI key
BJPLXUGLINHILT-UHFFFAOYSA-N
storage temp.
0-5C
Purity
95%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Yellow Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=8086
MSDS
http://www.chemcia.com/product_fl02.asp?id=8086
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.