CH4102151057
ChemBridge Corporation
(1,1-dimethyl-2-piperidin-1-ylethyl)amine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1,1-dimethyl-2-(1-piperidinyl)ethylamine
Inchi Code
1S/C9H20N2/c1-9(2,10)8-11-6-4-3-5-7-11/h3-8,10H2,1-2H3
InChI key
IMBNEKOMEDCTCL-UHFFFAOYSA-N
storage temp.
-20C
Purity
95%
Country of Origin
KZ
Shipping Temperature
RT
Physical Form
liquid
Product Link
https://www.hit2lead.com/building-blocks/4014539
MSDS
https://www.hit2lead.com/sds/4014539_sds.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.