Skip to Content
Merck
All Photos(1)

Documents

S-123

Supelco

AB-FUBINACA metabolite 2A solution

100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19FN4O4
Molecular Weight:
398.39
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−70°C

General description

AB-FUBINACA metabolite 2A is a major metabolite of AB-FUBINACA, a potent cannabinoid receptor agonist sold as a synthetic cannabinoid in Spice herbal blends. This certified reference solution is suited for use in synthetic cannabinoid testing methods by LC/MS o GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service