Skip to Content
Merck
All Photos(1)

Key Documents

N-114

Supelco

Norcarfentanil oxalate oxalate salt solution

100 μg/mL in methanol (as a free base), certified reference material, ampule of 0.5 mL, Cerilliant®

Synonym(s):

Norcarfentanil Oxalate Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 120.00

CHF 120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CHF 120.00

About This Item

Empirical Formula (Hill Notation):
C16H22N2O3 · C2H2O4
Molecular Weight:
380.39
UNSPSC Code:
41116107
NACRES:
NA.24

CHF 120.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as a free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for norcarfentanil LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Norcarfentanil is a major urinary metabolite of carfentanyl, a potent synthetic opioid analgesic drug structurally related to fentanyl. It is used in combination with other opioids and analgesics to induce or supplement general anesthesia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service