Skip to Content
Merck
All Photos(1)

Key Documents

L-039

Supelco

alpha-Linolenic acid

1.0 mg/mL in ethanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
CHF 200.00

CHF 200.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
CHF 200.00

About This Item

Empirical Formula (Hill Notation):
C18H30O2
CAS Number:
Molecular Weight:
278.43
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.24

CHF 200.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in ethanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC(=O)CCCCCCC\C=C/C\C=C/C\C=C/CC

InChI

1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10H,2,5,8,11-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-

InChI key

DTOSIQBPPRVQHS-PDBXOOCHSA-N

General description

a-Linolenic acid, also known as ALA, is an omega-3 fatty acid essential for human health and well-being. Measurement of ALA levels in serum is useful for evaluating essential fatty acid deficiency (EFAD), malabsorption and ALA treatment response in patients with EFAD. This certified solution standard is suitable for use as starting material in calibrators and controls for GC/MS fatty acid testing methods in clinical and diagnostic testing of ALA in patient samples.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service