Skip to Content
Merck
All Photos(1)

Key Documents

C-260

Supelco

Cannabivarinic acid (CBVA) solution

1 mg/mL (1% DIPEA and 0.05% Ascorbic Acid in Acetonitrile), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
CHF 183.00

CHF 183.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
CHF 183.00

About This Item

Empirical Formula (Hill Notation):
C20H22O4
CAS Number:
Molecular Weight:
326.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CHF 183.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (1% DIPEA and 0.05% Ascorbic Acid in Acetonitrile)

format

single component solution

storage temp.

−70°C

SMILES string

OC1=C(C(O)=O)C(CCC)=CC2=C1C3=C(C(C)(C)O2)C=CC(C)=C3

InChI

1S/C20H22O4/c1-5-6-12-10-15-17(18(21)16(12)19(22)23)13-9-11(2)7-8-14(13)20(3,4)24-15/h7-10,21H,5-6H2,1-4H3,(H,22,23)

InChI key

UTVPXAFTOXQUFY-UHFFFAOYSA-N

General description

Cannabivarinic acid (CBVA) is a natural phytocannabinoid. This certified Snap-N-Spike® solution is suitable as starting material in preparation of calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology, pharmaceutical research to cannabis testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service