A-144
Acylcarnitines Mix 2 (C4,iC4,C5,iC5,C6,C8,C10,C12,C14,C18) solution
100-200 μg/mL (90:10 Methanol: 1% aqueous formic acid), certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
Acylcarnitines Mix 2 Solution
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
composition
Butyryl-L-carnitine, 100 μg/mL
Decanoyl-L-carnitine, 100 μg/mL
Hexanoyl-L-carnitine, 100 μg/mL
Isobutyryl-L-carnitine, 100 μg/mL
Isovaleryl-L-carnitine, 100 μg/mL
Lauroyl-L-carnitine, 100 μg/mL
Myristoyl-L-carnitine, 100 μg/mL
Octanoyl-L-carnitine, 100 μg/mL
Stearoyl-L-carnitine, 200 μg/mL
Valeryl-L-carnitine, 100 μg/mL
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100-200 μg/mL (90:10 Methanol: 1% aqueous formic acid)
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
clinical testing
format
multi-component solution
storage temp.
−70°C
General description
Application
- Metabolic Disorder Biomarker Development: Acylcarnitines are crucial in diagnosing very long-chain acyl-CoA dehydrogenase deficiency. This research utilizes acylcarnitine profiles to improve understanding and treatment approaches for metabolic disorders, employing mass spectrometry metabolites for precise analysis (Bleeker JC et al., 2020).
- High-Purity Acylcarnitine Standards in Research: The study demonstrate the simultaneous extraction of metabolomes and lipidomes, including acylcarnitines, using ultra-high performance liquid chromatography/mass spectrometry. This methodology supports high-precision studies in fatty acid oxidation and metabolic assay development (Chen S et al., 2013).
- Application in Inborn Errors of Metabolism Assay: The research investigates how dietary bioactive polyphenols affect the mitochondrial carnitine acylcarnitine carrier, showcasing the utility of acylcarnitines in studying the impact of natural compounds on metabolic pathways, vital for understanding inborn errors of metabolism (Console L et al., 2019).
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
In this article we provide a complete workflow of quantification and identification of acylcarnitines in dried blood spots (DBS) using stable isotope-labeled (SIL) acylcarnitines CRMs by FIA-MS/MS.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service