Skip to Content
Merck
All Photos(1)

Key Documents

LM6002

Avanti

Cer/Sph Mixture I

Avanti Research - A Croda Brand LM6002, ethanol solution

Synonym(s):

Cer/Sph Mixture I, Ceramide/Sphingoid Internal Standard Mixture I

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 384.00

CHF 384.00


Estimated to ship on23 March 2025


Request a Bulk Order

Select a Size

Change View
1 EA
CHF 384.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

CHF 384.00


Estimated to ship on23 March 2025


Request a Bulk Order

form

ethanol solution

packaging

pkg of 1 × 1 mL (LM6002-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM6002

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

shipped in

dry ice

storage temp.

−20°C

Application

Cer/Sph Mixture I has been used:
  • as an internal standard in sphingolipid profiling[1]
  • in the preparation of ceramide-phosphoethanolamine extracts[2]
  • for the sphingolipid extraction[3]

Packaging

2 mL Amber Glass Sealed Ampule (LM6002-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeted lipid profiling discovers plasma biomarkers of acute brain injury
Sheth SA, et al.
PLoS ONE, 10(6), e0129735-e0129735 (2015)
Quantitation of ceramide phosphorylethanolamines containing saturated and unsaturated sphingoid base cores
Masood MA, et al.
Analytical Biochemistry, 400(2), 259-269 (2010)
Sally Chiu et al.
Nutrients, 12(2) (2020-02-09)
Soda consumption in adolescents has been linked to poorer metabolic outcomes. We tested whether replacing soda with reduced fat milk would improve features of atherogenic dyslipidemia and other cardiometabolic risk factors. Thirty overweight and obese adolescent boys who were habitual
A role for ceramides, but not sphingomyelins, as antagonists of insulin signaling and mitochondrial metabolism in C2C12 myotubes
Park M, et al.
The Journal of Biological Chemistry, 291(46), 23978-23988 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service