Skip to Content
Merck
All Photos(1)

Documents

900602P

Avanti

Photoclick C6 Ceramide

Avanti Research - A Croda Brand

Synonym(s):

N-((2S,3R,E)-1,3-dihydroxy-13-(3-(pent-4-yn-1-yl)-3H-diazirin-3-yl)tridec-4-en-2-yl)hexanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H43N3O3
CAS Number:
Molecular Weight:
433.63
UNSPSC Code:
12352211
NACRES:
NA.12

Assay

>99% (TLC)

form

powder

packaging

package of 1 × 250 μg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (900602P-250UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service