Skip to Content
Merck
All Photos(1)

Key Documents

900111P

Avanti

15β-PGF1α

powder

Synonym(s):

Prostaglandin F1α(15 beta epimer)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 148.00

CHF 148.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
CHF 148.00

About This Item

Empirical Formula (Hill Notation):
C20H36O5
CAS Number:
Molecular Weight:
356.50
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 148.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](O)CCCCC)[C@H](O)C1

InChI

1S/C20H36O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-19,21-23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16-,17-,18+,19-/m1/s1

InChI key

DZUXGQBLFALXCR-HBYHEDERSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (900111P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service