Skip to Content
Merck
All Photos(1)

Key Documents

880342P

Avanti

16:0-16:0(acrylate) PC

Avanti Research - A Croda Brand 880342P, powder

Synonym(s):

1-palmitoyl-2-[16-(acryloyloxy)palmitoyl]-sn-glycero-3-phosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H82NO10P
CAS Number:
Molecular Weight:
804.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (880342P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880342P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)C)=O)(OC(CCCCCCCCCCCCCCCOC(C=C)=O)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C43H82NO10P/c1-6-8-9-10-11-12-13-15-18-21-24-27-30-33-42(46)51-38-40(39-53-55(48,49)52-37-35-44(3,4)5)54-43(47)34-31-28-25-22-19-16-14-17-20-23-26-29-32-36-50-41(45)7-2/h7,40H,2,6,8-39H2,1,3-5H3/t40-/m1/s1

InChI key

RSMUYMLRJLXQQK-RRHRGVEJSA-N

Application

16:0-16:0(acrylate) PC or 1-palmitoyl-2-[16-(acryloyloxy)palmitoyl]-sn-glycero-3-phosphorylcholine is suitable for use in preparing functionalized glass surfaces to study colloidal particles interaction.

Packaging

5 mL Amber Glass Screw Cap Vial (880342P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Chon et al.
Journal of biomaterials science. Polymer edition, 10(1), 95-107 (1999-03-26)
The significance of molecular design methodologies based upon membrane-mimetic systems lies in the ability to engineer robust materials of varying geometry with a high degree of reproducibility and molecular control over surface order and chemistry. However, non-covalently associated assemblies, in
Interaction of colloidal particles with biologically relevant complex surfaces
Hernandez MJ, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 580, 123778-123778 (2019)
Patrick Drücker et al.
Langmuir : the ACS journal of surfaces and colloids, 30(49), 14877-14886 (2014-11-22)
Solid-supported lipid bilayers (SLBs) mimicking a biological membrane are commonly used to investigate lipid-lipid or lipid-protein interactions. Simple binary or ternary lipid systems are well established, whereas more complex model membranes containing biologically important signaling lipids such as phosphatidylinositol-4,5-bisphosphate (PI(4,5)P2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service