Skip to Content
Merck
All Photos(1)

Key Documents

870750P

Avanti

10:0 (3-Oxo) CoA

Avanti Research - A Croda Brand 870750P, powder

Synonym(s):

3-oxo decanoyl Coenzyme A

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 264.00

CHF 264.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
CHF 264.00

About This Item

Empirical Formula (Hill Notation):
C31H61N10O18P3S
CAS Number:
Molecular Weight:
986.86
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 264.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (870750P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870750P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CC(CCCCCCC)=O)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C31H52N7O18P3S/c1-4-5-6-7-8-9-19(39)14-22(41)60-13-12-33-21(40)10-11-34-29(44)26(43)31(2,3)16-53-59(50,51)56-58(48,49)52-15-20-25(55-57(45,46)47)24(42)30(54-20)38-18-37-23-27(32)35-17-36-28(23)38/h17-18,20,24-26,30,42-43H,4-16H2,1-3H3,(H,33,40)(H,34,44)(H,48,49)(H,50,51)(H2,32,35,36)(H2,45,46,47)

InChI key

AZCVXMAPLHSIKY-UHFFFAOYSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (870750P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service