Skip to Content
Merck
All Photos(1)

Documents

870729P

Avanti

25:0 Coenzyme A

Avanti Research - A Croda Brand 870729P, powder

Synonym(s):

pentacosanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H93N10O17P3S
CAS Number:
Molecular Weight:
1183.27
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870729P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870729P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

General description

25:0 Coenzyme A or pentacosanoyl coenzyme A is a long chain fatty acyl-CoA. Fatty acyl-CoAs is produced from de-novo fatty acid biosynthesis and the reprocessing of pre-existing fatty acids.
25:0 Coenzyme A, also known as pentacosanoyl coenzyme A, is a coenzyme derivative of pentacosanoic acid.

Application

25:0 Coenzyme A has been used as a constituent of the internal standard mixture in liquid chromatography-electrospray ionization tandem mass spectrometry (LC/ESI-MS/MS).
25:0 Coenzyme A or pentacosanoyl Coenzyme A has been used as a substrate for 1-acylglycerol-3-phosphate-O-acyltransferase 11 (AGPAT11) enzyme. It has also been used as an internal standard to quantify fatty acyl CoA extracted from human embryonic kidney 293 cells.

Biochem/physiol Actions

Fatty acyl-CoA plays a vital role in various cellular processes including, energy metabolism, the biosynthesis and recycling of complex lipids, the posttranslational modification of proteins and the regulation of gene expression. In addition, it facilitates most of the fatty acid modification reactions. 25:0 coenzyme A might function as a precursor of sphingolipids. Alterations in the fatty acyl-CoA metabolism leads to obesity, cardiovascular disease, diabetes mellitis and cancer.

Packaging

5 mL Amber Glass Screw Cap Vial (870729P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Factors to consider in using [UC] palmitate for analysis of sphingolipid biosynthesis by tandem mass spectrometry
Haynes CA, et al.
Journal of Lipid Research, 52(8), 1583-1594 (2011)
Quantitation of fatty acyl-coenzyme As in mammalian cells by liquid chromatography-electrospray ionization tandem mass spectrometry
Haynes CA, et al.
Journal of Lipid Research, 49(5), 1113-1125 (2008)
Fatty acid oxidation and control of food intake
Leonhardt M and Langhans W
Physiology & Behavior, 83(4), 645-651 (2004)
Christopher A Haynes et al.
Journal of lipid research, 52(8), 1583-1594 (2011-05-19)
This study describes the use of a stable-isotope labeled precursor ([U-¹³C]palmitate) to analyze de novo sphingolipid biosynthesis by tandem mass spectrometry. It also describes factors to consider in interpreting the data, including the isotope's location (¹³C appears in three isotopomers
Role of fatty acid binding proteins and long chain fatty acids in modulating nuclear receptors and gene transcription
Schroeder F, et al.
Lipids, 43(1), 1-17 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service