Skip to Content
Merck
All Photos(1)

Key Documents

870650P

Avanti

Trans-FAAzo-4

Avanti Research - A Croda Brand, powder

Synonym(s):

4-Butyl-Azo-4:0-Acid-1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 169.00
25 MG
CHF 641.00

CHF 169.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CHF 169.00
25 MG
CHF 641.00

About This Item

Empirical Formula (Hill Notation):
C20H24N2O2
CAS Number:
Molecular Weight:
324.42
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 169.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

1 ea of 25 mg (870650P-25mg)
1 ea of 5 mg (870650P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

photoswitchable lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC1=CC=C(/N=N/C2=CC=C(CCCC(O)=O)C=C2)C=C1

InChI

1S/C20H24N2O2/c1-2-3-5-16-8-12-18(13-9-16)21-22-19-14-10-17(11-15-19)6-4-7-20(23)24/h8-15H,2-7H2,1H3,(H,23,24)/b22-21+

InChI key

NZZOOAGSNMPTTQ-QURGRASLSA-N

General description

FAAzos are photoswitchable fatty acids which mimics highly unsaturated fatty acids such as arachidonic acid. FAAzo-4 is an azobenzene derivative.[1]

Biochem/physiol Actions

FAAzos are potential modular building blocks for synthetic incorporation into more elaborate photolipids.[1] FAAzo has the potential for the optical control of G-protein coupled receptor (GPCR).[2]

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical control of GPR40 signalling in pancreatic beta-cells
Frank JA, et al.
Chemical Science, 8(11), 7604-7610 (2017)
Photoswitchable diacylglycerols enable optical control of protein kinase C
Frank JA, et al.
Nature Chemical Biology, 12(9), 755-755 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service