Skip to Content
Merck
All Photos(1)

Key Documents

870222P

Avanti

18:1 Succinyl PE

Avanti Research - A Croda Brand 870222P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(succinyl) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CHF 181.00
200 MG
CHF 689.00

CHF 181.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
CHF 181.00
200 MG
CHF 689.00

About This Item

Empirical Formula (Hill Notation):
C45H81NO11PNa
CAS Number:
Molecular Weight:
866.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 181.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 2 × 100 mg (870222P-200mg)
pkg of 1 × 25 mg (870222P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870222P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCC(O)=O)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C45H82NO11P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-44(50)54-39-41(40-56-58(52,53)55-38-37-46-42(47)35-36-43(48)49)57-45(51)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,41H,3-16,21-40H2,1-2H3,(H,46,47)(H,48,49)(H,52,53);/q;+1/p-1/b19-17-,20-18-;/t41-;/m1./s1

InChI key

KKEGOYIHHIJUOK-NFCDTPQPSA-M

General description

18:1 Succinyl PE , also known as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(succinyl) (N-succinyl DPPE), is a head group modified functionalized lipid. Functionalized lipids perform the role of reporter molecules.[1] N-succinyl DPPE in combination with poly(vinyl pyrrolidone) is used in preparation of new drug delivery systems.[2]

Packaging

5 mL Clear Glass Sealed Ampule (870222P-200mg)
5 mL Clear Glass Sealed Ampule (870222P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (vinyl pyrrolidone) conjugated lipid system for the hydrophobic drug delivery
Lee HY, et al.
Macromolecular Research, 15(6), 547-552 (2007)
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service