Skip to Content
Merck
All Photos(1)

Key Documents

860832P

Avanti

C6(6-azido) LacCer

Avanti Research - A Croda Brand 860832P, powder

Synonym(s):

D-lactosyl-β-1,1′ N-(6"-azidohexanoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 287.00

CHF 287.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 287.00

About This Item

Empirical Formula (Hill Notation):
C36H66 N4O13
CAS Number:
Molecular Weight:
762.93
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 287.00


Estimated to ship on24 May 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860832P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860832P

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCN=[N+]=[N-])=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

InChI

1S/C36H66N4O13/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-25(43)24(39-28(44)19-16-14-17-20-38-40-37)23-50-35-33(49)31(47)34(27(22-42)52-35)53-36-32(48)30(46)29(45)26(21-41)51-36/h15,18,24-27,29-36,41-43,45-49H,2-14,16-17,19-23H2,1H3,(H,39,44)/b18-15+/t24-,25+,2

InChI key

DHYANRUMYSYKQW-KONZMJRYSA-N

General description

C6(6-azido) LacCer or D-lactosyl-β-1,1′ N-(6"-azidohexanoyl)-D-erythro-sphingosine, is a head group modified lipid containing an omega-terminal azide. This click reagent with the terminal azide group can be used in a highly specific linking reaction with alkyne-containing reagents, known as ‘click chemistry′, in the presence of a copper (Cu)-containing catalyst.

Packaging

5 mL Amber Glass Screw Cap Vial (860832P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service