Skip to Content
Merck
All Photos(2)

Key Documents

860649P

Avanti

C24 Ceramide (d17:1/24:0)

Avanti Research - A Croda Brand 860649P, powder

Synonym(s):

N-lignoceroyl-D-erythro-sphingosine (C17 base)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 695.00

CHF 695.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CHF 695.00

About This Item

Empirical Formula (Hill Notation):
C41H81NO3
CAS Number:
Molecular Weight:
636.09
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 695.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (860649P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860649P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCC

InChI

1S/C41H81NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-25-27-29-31-33-35-37-41(45)42-39(38-43)40(44)36-34-32-30-28-26-16-14-12-10-8-6-4-2/h34,36,39-40,43-44H,3-33,35,37-38H2,1-2H3,(H,42,45)/b36-34+/t39-,40+/m0/s1

InChI key

OWQPALVMZWREMV-PUYNVXOJSA-N

General description

C24 Ceramide (d17:1/24:0) is a synthetic ceramide derived from a C17 sphingosine with lignoceric acid.[1] Ceramide synthase 2 is responsible for the synthesis of C24 ceramide.[2]

Application

C24 Ceramide (d17:1/24:0) has been used as an internal standard to quantify ceramide content in both subcutaneous and visceral adipose tissue by ultraperformance liquid chromatography - tandem mass spectrometery (UPLC-MS/MS).[3]

Biochem/physiol Actions

C24 Ceramide (d17:1/24:0) plays a vital role in inducing cell death.[1] Altered levels of C24 Ceramide might be a potential indication in tumor monitoring.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860649P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka U Blachnio-Zabielska et al.
International journal of molecular sciences, 19(12) (2018-12-14)
Ceramide accumulation in muscle and in liver is implicated in the induction of insulin resistance. Much less in known about the role of ceramide in adipose tissue. The aim of the present study was to elucidate the role of ceramide
Luisa Coderch et al.
American journal of clinical dermatology, 4(2), 107-129 (2003-01-30)
Ceramides are the major lipid constituent of lamellar sheets present in the intercellular spaces of the stratum corneum. These lamellar sheets are thought to provide the barrier property of the epidermis. It is generally accepted that the intercellular lipid domain
Ling Chen et al.
Oncology reports, 34(1), 447-454 (2015-05-16)
Endocannabinoids and ceramides have demonstrated growth inhibition, cell death induction and pro-apoptotic activity in cancer research. In the present study, we describe the profiles of two major endocannabinoids, ceramides, free fatty acids and relevant metabolic enzymes in 47 pairs of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service