Skip to Content
Merck
All Photos(1)

Documents

860597P

Avanti

C24:1 Lactosyl(β) Ceramide (d18:1/24:1)

Avanti Research - A Croda Brand 860597P, powder

Synonym(s):

D-lactosyl-β1-1′-N-nervonoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H101NO13
CAS Number:
Molecular Weight:
972.38
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860597P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860597P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

General description

Lactosylceramide (LacCer) is a glycosphingolipid, which has a hydrophobic ceramide lipid and a hydrophilic sugar moiety. It is present mainly in the nervous system and myelin sheath.

Biochem/physiol Actions

Lactosylceramide (LacCer) is a structural and signaling lipid. It aids microbial attachment and also functions as an adhesion receptor for pathogens. LacCer is essential for signal transduction and cell adhesion. It controls cell proliferation and apoptosis. LacCer is implicated in cellular stress response and microbial pathogenesis. It facilitates innate immune responses and host-tissue pathogen interactions.

Packaging

5 mL Amber Glass Screw Cap Vial (860597P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry, Biochemistry, and Biology of 1-3 Beta Glucans and Related Polysaccharide, 191, 106-114 (2009)
Jarod A Fincher et al.
Scientific reports, 9(1), 17508-17508 (2019-11-27)
Neutral lipids have been implicated in a host of potentially debilitating human diseases, such as heart disease, type-2 diabetes, and metabolic syndrome. Matrix-assisted laser desorption ionization (MALDI), the method-of-choice for mass spectrometry imaging (MSI), has led to remarkable success in
Daniela Couto et al.
Chemistry and physics of lipids, 191, 106-114 (2015-09-01)
Galactosylceramide (GalCer) and lactosylceramide (LacCer) are structural and signaling lipids, playing important roles in signal transduction and cell adhesion. They are especially abundant in the nervous system and in important components of the myelin sheath. Although neurodegenerative disorders are associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service