Skip to Content
Merck
All Photos(1)

Key Documents

860575P

Avanti

C8 L-threo-Lactosyl(β) Ceramide (d18:1/8:0)

Avanti Research - A Croda Brand 860575P, powder

Synonym(s):

D-lactosyl-β1-1′-N-octanoyl-L-threo-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 159.00

CHF 159.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
CHF 159.00

About This Item

Empirical Formula (Hill Notation):
C38H71NO13
CAS Number:
Molecular Weight:
749.97
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 159.00


Estimated to ship on31 May 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860575P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860575P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@](O)([H])[C@@]([H])(NC(CCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

InChI key

JUCWRRXMPGRQOG-CKTHHAJGSA-N

General description

C8 L-threo-Lactosyl(β) Ceramide (d18:1/8:0) is a synthetic glycosphingolipid (GSL) with nonnatural stereochemistry.

Application

C8 L-threo-Lactosyl(β) Ceramide (d18:1/8:0) may be used to disrupt brain-derived neurotropic factor (BDNF)-induced integrin clustering.[1]

Biochem/physiol Actions

C8 L-threo-Lactosyl(β) Ceramide (d18:1/8:0) is a caveolar uptake inhibitor, which blocks SV40 virus infection. It also prevents E1-integrin activation and downstream signaling.[1] C8 L-threo-Lactosyl(β) Ceramide (d18:1/8:0) can deter the aggregation of lipids and proteins into glycosphingolipids (GSLs) and cholesterol-enriched microdomains (rafts) at the plasma membrane (PM).

Packaging

5 mL Amber Glass Screw Cap Vial (860575P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bidirectional remodeling of beta1-integrin adhesions during chemotropic regulation of nerve growth
Carlstrom LP, et al.
BMC biology, 9(1), 82-82 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service