Skip to Content
Merck
All Photos(2)

Documents

860399C

Avanti

16:0-d31-18:1 PC

1-palmitoyl-d31-2-oleoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

110918

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H51NO8PD31
CAS Number:
Molecular Weight:
791.27
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (860399C-10mg)
pkg of 1 × 2.5 mL (860399C-25mg)
pkg of 1 × 4 mL (860399C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860399C

concentration

10 mg/mL (860399C-10mg)
10 mg/mL (860399C-25mg)
25 mg/mL (860399C-100mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)C)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1/i2D3,7D2,9D2,11D2,13D2,15D2,17D2,19D2,22D2,24D2,26D2,28D2,30D2,32D2,34D2

InChI key

WTJKGGKOPKCXLL-KHCKXXADSA-N

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0-d31-18:1 PC may be used in the preparation of mitochondrial mimetic liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (860399C-100mg)
5 mL Clear Glass Sealed Ampule (860399C-10mg)
5 mL Clear Glass Sealed Ampule (860399C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delineating the rules for structural adaptation of membrane-associated proteins to evolutionary changes in membrane lipidome
Makarova M, et al.
Current Biology (2020)
Surface-binding to cardiolipin nanodomains triggers cytochrome c pro-apoptotic peroxidase activity via localized dynamics
Li M, et al.
Structure, 27(5), 806-815 (2019)
Rita Touti et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 151, 18-31 (2020-04-11)
Multi-lamellar liposomes (MLLs), prepared by shearing a lamellar phase composed of lipids (phosphatidylcholine) and surfactant (Tween 80®), were designed to control their size, charge and elasticity, the key parameters known to influence liposomes penetration through skin. Their size was tuned
Kotaro Hama et al.
Journal of lipid research, 61(4), 523-536 (2020-02-23)
X-linked adrenoleukodystrophy (X-ALD) is an inherited disorder caused by deleterious mutations in the ABCD1 gene. The ABCD1 protein transports very long-chain FAs (VLCFAs) from the cytosol into the peroxisome where the VLCFAs are degraded through β-oxidation. ABCD1 dysfunction leads to
Alessandra Luchini et al.
Analytical chemistry, 92(1), 1081-1088 (2019-11-27)
In vitro characterization of membrane proteins requires experimental approaches providing mimics of the microenvironment that proteins encounter in native membranes. In this context, supported lipid bilayers provide a suitable platform to investigate membrane proteins by a broad range of surface-sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service