Skip to Content
Merck
All Photos(2)

Key Documents

860342P

Avanti

14:0 PC-d9

1,2-dimyristoyl-sn-glycero-3-phosphocholine-N,N,N-trimethyl-d9, powder

Synonym(s):

[(2R)-2,3-di(tetradecanoyloxy)propyl] 2-[tris(trideuteriomethyl)azaniumyl]ethyl phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 1’230.00

CHF 1’230.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
CHF 1’230.00

About This Item

Empirical Formula (Hill Notation):
C36H63NO8PD9
CAS Number:
Molecular Weight:
686.99
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

CHF 1’230.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860342P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860342P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O

InChI

1S/C36H72NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-35(38)42-32-34(33-44-46(40,41)43-31-30-37(3,4)5)45-36(39)29-27-25-23-21-19-17-15-13-11-9-7-2/h34H,6-33H2,1-5H3/t34-/m1/s1/i3D3,4D3,5D3

InChI key

CITHEXJVPOWHKC-LNEGZTHBSA-N

General description

14:0 PC-d9 is a deuterated phosphocholine (PC) wherein nine protons of dimyristoyl are replaced by deuterium.

Packaging

5 mL Amber Glass Screw Cap Vial (860342P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of materials chemistry. B (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service