Skip to Content
Merck
All Photos(1)

Key Documents

857523P

Avanti

N-Tetradecanoyl-L-homoserine lactone

Avanti Research - A Croda Brand

Synonym(s):

(S)-N-(2-Oxotetrahydrofuran-3-yl)tetradecanamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 187.00
50 MG
CHF 560.00

CHF 187.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CHF 187.00
50 MG
CHF 560.00

About This Item

Empirical Formula (Hill Notation):
C18H33NO3
CAS Number:
Molecular Weight:
311.46
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 187.00


Estimated to ship on31 May 2025


Request a Bulk Order

description

(S)-N-(2-Oxotetrahydrofuran-3-yl)tetradecanamide

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (857523P-10MG)
pkg of 1 × 50 mg (857523P-50MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bacterial lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C1OCC[C@@H]1NC(CCCCCCCCCCCCC)=O

InChI

1S/C18H33NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-17(20)19-16-14-15-22-18(16)21/h16H,2-15H2,1H3,(H,19,20)/t16-/m0/s1

InChI key

ZQAYHOXXVBVXPZ-INIZCTEOSA-N

General description

N-Tetradecanoyl-L-homoserine lactone/C14-HSL is a N-acylhomoserine lactone (HSL). C14-HSL is a non-polar compound.[1] All HSLs possess a homoserine lactone moiety and a fatty acyl group with varying carbon length ranging from 4 to 14 carbons. They also have either 3-oxo, 3-hydroxyl, or fully reduced methylene groups at the C-3 position.[2]

Biochem/physiol Actions

N-acylhomoserine lactones derivatives (HSLs) are produced and utilized by Gram negative bacteria as signaling molecules, to invade and colonize higher organisms.[2] It is a quorum sensing signaling molecule commonly modulates cell density-dependent regulatory mechanisms.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (857523P-10MG and 857523P-50MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occurrence of N-acyl-L-homoserine lactones in extracts of some Gram-negative bacteria evaluated by gas chromatography-mass spectrometry.
Cataldi TRI, et al.
Analytical biochemistry, 361(2), 226-235 (2007)
Acyl homoserine lactone-based quorum sensing in a methanogenic archaeon.
Zhang G, et al.
The ISME Journal, 6(7), 1336-1336 (2012)
The hydrolysis of unsubstituted N-acylhomoserine lactones to their homoserine metabolites: Analytical approaches using ultra performance liquid chromatography.
Englmann M, et al.
Journal of Chromatography A, 1160(1-2), 184-193 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service