Skip to Content
Merck
All Photos(2)

Documents

855176P

Avanti

07:0 Lyso PC

1-heptanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-heptanoyl-sn-glycero-3-phosphocholine; PC(7:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H32NO7P
CAS Number:
Molecular Weight:
369.39
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855176P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855176P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCC)=O

General description

07:0 Lyso PC, a natural lysophosphatidylcholine (Lyso-PC), is a bioactive proinflammatory lipid, produced on pathological response. It is a component of blood plasma, vascular tissue and lipoproteins.

Biochem/physiol Actions

Lysophosphatidylcholine (Lyso-PC) augments the expression of genes for growth factors and cellular adhesion molecules. It is linked to inflammatory diseases and atherosclerosis.

Packaging

5 mL Clear Glass Sealed Ampule (855176P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a UHPLC-ESI-MS/MS method for the simultaneous quantification of mammal lysophosphatidylcholines and lysophosphatidylethanolamines in serum
Suarez-Garcia S, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 1055, 86-97 (2017)
Lysophosphatidylcholine stimulates the release of arachidonic acid in human endothelial cells
Wong JT, et al.
Test, 273(12), 6830-6836 (1998)
Role of lysophosphatidylcholine (LPC) in atherosclerosis
Matsumoto T, et al.
Current Medicinal Chemistry, 14(30), 3209-3220 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service