Skip to Content
Merck
All Photos(2)

Key Documents

840860C

Avanti

16:0-22:6 PA

1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1-hexadecanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphate (sodium salt); PA(16:0/22:6(4Z,7Z,10Z,13Z,16Z,19Z)); 110617

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CHF 610.00

CHF 610.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
CHF 610.00

About This Item

Empirical Formula (Hill Notation):
C41H68O8PNa
CAS Number:
Molecular Weight:
742.94
UNSPSC Code:
51191904
NACRES:
NA.25

CHF 610.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840860C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840860C

concentration

10 mg/mL (840860C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C41H69O8P.Na/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-24-26-28-30-32-34-36-41(43)49-39(38-48-50(44,45)46)37-47-40(42)35-33-31-29-27-25-23-16-14-12-10-8-6-4-2;/h5,7,11,13,17-18,20-21,24,26,30,32,39H,3-4,6,8-10,12,14-16,19,22-23,25,27-29,31,33-38H2,1-2H3,(H2,44,45,46);/q;+1/p-1/b7-5-,13-11-,18-17-,21-20-,26-24-,32-30-;/t39-;/m1./s1

InChI key

JVHJPADGSWMGQX-KQTQLPLBSA-M

General description

16:0-22:6 PA or 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphate (sodium) is a sodium salt of phosphatidic acid, with substitution of palmitic acid and docosahexaenic acid at the sn-1 and sn-2 positions respectively, in the glycerol backbone.

Application

16:0-22:6 PA or 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphate (sodium salt) has been used as a calibration standard for lipidomic analysis.[1]

Packaging

5 mL Clear Glass Sealed Ampule (840860C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ségolène Hescot et al.
Oncotarget, 8(66), 109924-109940 (2018-01-05)
Mitotane (o,p'DDD), the most effective drug in adrenocortical carcinoma, concentrates into the mitochondria and impacts mitochondrial functions. To address the molecular mechanisms of mitotane action and to identify its potential target, metabolomic and lipidomic approaches as well as imaging analyses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service