Skip to Content
Merck
All Photos(1)

Key Documents

790413P

Avanti

Coag Reagent II

Avanti Research - A Croda Brand 790413P, powder

Synonym(s):

Coag Reagent II, Coagulation Reagent II, contains DOPC:DOPS (7:3, w/w)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CHF 340.00

CHF 340.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
CHF 340.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

CHF 340.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 2 × 100 mg (790413P-200mg)
pkg of 1 × 25 mg (790413P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790413P

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Coag Reagent II is a lipid blend.

Packaging

5 mL Clear Glass Sealed Ampule (790413P-200mg)
5 mL Clear Glass Sealed Ampule (790413P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service