Skip to Content
Merck
All Photos(1)

Key Documents

700227P

Avanti

Cholenic acid

Avanti Research - A Croda Brand

Synonym(s):

3β-Hydroxy-5-cholenic acid, 3β-hydroxychol-5-en-24-oic acid, 3-hydroxy-5-cholenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 256.00

CHF 256.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
CHF 256.00

About This Item

Empirical Formula (Hill Notation):
C24H38O3
CAS Number:
Molecular Weight:
374.56
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 256.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 100 mg

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bile acids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC1CCC2(C3C(C4C(C(CC4)C(CCC(=O)O)C)(CC3)C)CC=C2C1)C

InChI

1S/C24H38O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h5,15,17-21,25H,4,6-14H2,1-3H3,(H,26,27)

InChI key

HIAJCGFYHIANNA-UHFFFAOYSA-N

General description

Cholenic acid is a monohydroxy bile acid.[1]

Biochem/physiol Actions

Cholenic acid serves as a precursor for chenodeoxycholic acid.[1]

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N B Javitt et al.
The Journal of biological chemistry, 261(27), 12486-12489 (1986-09-25)
Metabolism of 3 beta-hydroxy-5-cholenoic acid to chenodeoxycholic acid has been found to occur in rabbits and humans, species that cannot 7 alpha-hydroxylate lithocholic acid. This novel pathway for chenodeoxycholic acid synthesis from 3 beta-hydroxy-5-cholenoic acid led to a reinvestigation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service