Skip to Content
Merck
All Photos(1)

Key Documents

700114P

Avanti

5α-7,24-cholestadiene

Avanti Research - A Croda Brand 700114P, powder

Synonym(s):

5α-cholesta-7,24-dien-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 404.00

CHF 404.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
CHF 404.00

About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CHF 404.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700114P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700114P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1C[C@H]2[C@@]([C@H]3CC[C@]4([C@@H](CC[C@@H]4[C@@H](CCC=C(C)C)C)C3=CC2)C)(CC1)C

InChI

1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,10,19-21,23-25,28H,6,8-9,11-17H2,1-5H3/t19-,20+,21+,23-,24+,25+,26+,27-/m1/s1

InChI key

PKEPPDGGTSZLBL-SKCNUYALSA-N

General description

Oxygenated derivatives of cholesterol (oxysterols) present a remarkably diverse profile of biological activities, including effects on sphingolipid metabolism, platelet aggregation, apoptosis, protein prenylation, and cholesterol homeostasis.

Packaging

5 mL Amber Glass Screw Cap Vial (700114P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service