Skip to Content
Merck
All Photos(2)

Key Documents

700026P

Avanti

3β-hydroxy-7-oxo-5-cholestenoic acid

Avanti Research - A Croda Brand

Synonym(s):

(25R)-cholest-5-en-26-oic acid, 3β-hydroxy-7-oxo

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42O4
CAS Number:
Molecular Weight:
430.62
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700026P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Cholestenoic acids are cholesterol metabolic intermediates and precursors to bile acids. 3β-hydroxy-7-oxo-5-cholestenoic acid is synthesized from 7β-hydroxycholesterol. The enzymes for synthesis of cholestenoic acids are majorly present in the central nervous system (CNS) of mammals. 3β-hydroxy-7-oxo-5-cholestenoic acid (3βH,7O-CA) is also synthesized from 26-hydroxy-7-oxocholesterol by the action of the enzyme cytochrome P450 family 27 subfamily A member 1 sterol 27-hydroxylase (CYP27A1).

Biochem/physiol Actions

3β-hydroxy-7-oxocholest-5-en-26-oic acid (3βH,7O-CA) acts as a ligand for liver X receptors (LXR) and activates them at micromolar concentration. 3βH,7O-CA favors the islet-1+ cells synthesis and islet-1–GFP expression. 3βH,7O-CA also mediates oculomotor neurons maturation and its conversion to 3β,7β-dihydroxycholest-5-en-(25S)26-oic acid (3β,7β-diHCA) is catalyzed by the enzyme hydroxysteroid 11-β dehydrogenase.

Packaging

5 mL Amber Glass Screw Cap Vial (700026P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholestenoic acids regulate motor neuron survival via liver X receptors
Theofilopoulos S, et al.
The Journal of Clinical Investigation, 124(11), 4829-4842 (2014)
Sterolomics in biology, biochemistry, medicine
Griffiths WJ and Wang Y
TrAC, Trends in Analytical Chemistry, 115280-115280 (2018)
Identification of unusual oxysterols and bile acids with 7-oxo or 3beta, 5alpha, 6beta-trihydroxy functions in human plasma by charge-tagging mass spectrometry with multistage fragmentation
Griffiths WJ, et al.
Journal of Lipid Research, 59(6), 1058-1070 (2018)
27-Hydroxylated Low Density Lipoprotein (LDL) Cholesterol Can Be Converted to 7alpha, 27-Dihydroxy-4-cholesten-3-one (Cytosterone) before Suppressing Cholesterol Production in Normal Human Fibroblasts EVIDENCE THAT AN ALTERED METABOLISM OF LDL CHOLESTEROL
Axelson M and Larsson O
The Journal of Biological Chemistry, 271(22), 12724-12736 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service