Skip to Content
Merck
All Photos(1)

Key Documents

ATC311765060

Atlantic Research Chemicals Ltd

4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
364.4
MDL number:

Iupac Name

4-[(benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)-2-piperazinecarboxylic acid

Inchi Code

1S/C18H24N2O6/c1-18(2,3)26-17(24)20-10-9-19(11-14(20)15(21)22)16(23)25-12-13-7-5-4-6-8-13/h4-8,14H,9-12H2,1-3H3,(H,21,22)

InChI key

SREPAMKILVVDSP-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=CA03801

MSDS

https://atlantic-chemicals.com/product/?code=CA03801


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library