Skip to Content
Merck
All Photos(1)

Key Documents

ATE443543364

AstaTech, Inc.

2-AZABICYCLO[2.2.2]OCTANE-2,6-DICARBOXYLIC ACID 2-TERTBUTYL ESTER

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
255.31
MDL number:

Iupac Name

2-(tert-butoxycarbonyl)-2-azabicyclo[2.2.2]octane-6-carboxylic acid

Inchi Code

1S/C13H21NO4/c1-13(2,3)18-12(17)14-7-8-4-5-10(14)9(6-8)11(15)16/h8-10H,4-7H2,1-3H3,(H,15,16)

InChI key

BICQNBQLGTZYCQ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Shipping Temperature

Normal

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=52506

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=37778


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library