ATE413072565
AstaTech, Inc.
1-ETHYL-4-(4-NITROPHENYL)PIPERAZINE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-ethyl-4-(4-nitrophenyl)piperazine
Inchi Code
1S/C12H17N3O2/c1-2-13-7-9-14(10-8-13)11-3-5-12(6-4-11)15(16)17/h3-6H,2,7-10H2,1H3
InChI key
XEWICNRVCQLKIG-UHFFFAOYSA-N
storage temp.
2-8 C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
Light-red to Brown Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=26743
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=23541
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service