Skip to Content
Merck
All Photos(1)

Key Documents

APO456794502

Apollo Scientific Ltd

3-(3-Bromophenyl)prop-2-yn-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
211.06
MDL number:

Iupac Name

3-(3-bromophenyl)-2-propyn-1-ol

Inchi Code

1S/C9H7BrO/c10-9-5-1-3-8(7-9)4-2-6-11/h1,3,5,7,11H,6H2

InChI key

UNSAJINGUOTTRA-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/3-3-bromophenylprop-2-yn-1-ol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-3-bromophenylprop-2-yn-1-ol


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library