Z743295
Julabo® adaptors
twin distributing with barbed fittings, for 8 mm i.d. tubing, pack of 1
Synonym(s):
barbed adaptor
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
packaging
pack of 1
manufacturer/tradename
Julabo 8970470
Looking for similar products? Visit Product Comparison Guide
General description
For use with Julabo® circulators, twin distributing with barbed fittings, for 8 mm i.d. tubing, pack of 1.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service