Key Documents
Z742448
MAGic Clamp magnetic clamp
holds 3000 mL Erlenmeyer flask
Synonym(s):
flask clamps, magnetic clamps
Select a Size
1 EA
CHF 125.00
CHF 125.00
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 125.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.73
CHF 125.00
Check Cart for Availability
feature
holds 3000 mL Erlenmeyer flask
max 4x3000ml flask
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific H1000-MR-3000
General description
The convenient MAGic Clam platform for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.
designed to be used with Benchmark Scientific H1010 shaking incubators and OrbiShaker XL orbital shakers
Legal Information
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)