Key Documents
Z740687
Sigma-Aldrich
Aldrich® Essentials filter flask, with side arm
capacity 2000 mL
Select a Size
4 EA
CHF 638.00
CHF 638.00
Check Cart for Availability
Select a Size
Change View
4 EA
CHF 638.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
CHF 638.00
Check Cart for Availability
material
borosilicate glass flask
flat bottom flask
Joint
joint stopper9
packaging
pkg of 4 ea
capacity
2000 mL
General description
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)