Key Documents
Z740554
Sigma-Aldrich
Aldrich® Essentials beaker, Griffin
low form, capacity 5 mL, ungraduated
Select a Size
12 EA
CHF 79.00
CHF 79.00
Check Cart for Availability
Select a Size
Change View
12 EA
CHF 79.00
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
CHF 79.00
Check Cart for Availability
material
borosilicate glass
feature
graduations: no
packaging
pkg of 12 ea
manufacturer/tradename
Sigma-Aldrich
capacity
5 mL
Related Categories
General description
Borosilicate glass, Griffin style, low form beakers with reference graduations and pour spout. Uniform, standard walls and flat bottom for even heating. Meet ASTM E960 designation.
Other Notes
Browse all glass beakers
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)